Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040419.3 | NP_001035509.1 | 712 | 127 | ||
GenBank mRNA | JU322643.1 | - | 676 | 127 | ||
JU473227.1 | - | 834 | 127 | |||
JV566863.1 | - | 712 | 127 | |||
JV721926.1 | - | 707 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |