Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119345.2 | XP_014974831.2 | 669 | 108 | ||
GenBank mRNA | JU327376.1 | - | 428 | 108 | ||
JV659249.1 | - | 400 | 108 | |||
JV667270.1 | - | 689 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |