Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261794.1 | NP_001248723.1 | 312 | 71 | ||
XM_015120247.2 | XP_014975733.1 | 597 | 71 | |||
GenBank mRNA | JU471957.1 | - | 312 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |