Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194172.3 | NP_001181101.1 | 375 | 152 | ||
XM_015145313.2 | XP_015000799.1 | 375 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |