Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839136.1 | XP_028694969.1 | 1246 | 147 | ||
GenBank mRNA | JU328466.1 | - | 324 | 147 | ||
JU477226.1 | - | 1142 | 147 | |||
JV071448.1 | - | 1047 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |