Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839136.1 | XP_028694969.1 | 1717 | 63 | ||
GenBank mRNA | JU328466.1 | - | 795 | 63 | ||
JU477226.1 | - | 1613 | 63 | |||
JV071448.1 | - | 1518 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |