Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111465.3 | XP_001111465.1 | 602 | 169 | ||
XM_015135655.2 | XP_014991141.1 | 599 | 169 | |||
XM_028835885.1 | XP_028691718.1 | 602 | 169 | |||
GenBank mRNA | JU493902.1 | - | 519 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |