Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087439.4 | XP_001087439.2 | 1128 | 157 | ||
XM_015120959.2 | XP_014976445.2 | 460 | 157 | |||
GenBank mRNA | JU477301.1 | - | 1116 | 157 | ||
JU477302.1 | - | 1116 | 157 | |||
JV048305.1 | - | 453 | 157 | |||
JV724005.1 | - | 778 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |