Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1LG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083599.2 | NP_001077068.1 | 534 | 117 | ||
XM_015117664.2 | XP_014973150.1 | 757 | 117 | |||
GenBank mRNA | EF444815.1 | - | 535 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |