Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194774.2 | NP_001181703.2 | 1431 | 145 | ||
XM_001111429.4 | XP_001111429.2 | 1262 | 145 | |||
XM_015111034.2 | XP_014966520.2 | 1466 | 145 | |||
XM_015111036.2 | XP_014966522.2 | 1776 | 145 | |||
XM_015111037.2 | XP_014966523.2 | 2100 | 145 | |||
XM_015111039.2 | XP_014966525.2 | 1397 | 145 | |||
XM_015111040.2 | XP_014966526.2 | 1693 | 145 | |||
XM_028830550.1 | XP_028686383.1 | 1639 | 145 | |||
XM_028830551.1 | XP_028686384.1 | 1118 | 145 | |||
GenBank mRNA | JV724118.1 | - | 676 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |