Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261606.1 | NP_001248535.1 | 206 | 70 | ||
XM_015128312.2 | XP_014983798.1 | 1401 | 70 | |||
GenBank mRNA | JU321908.1 | - | 707 | 70 | ||
JU473350.1 | - | 677 | 70 | |||
JV048326.1 | - | 206 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |