Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261583.1 | NP_001248512.1 | 784 | 132 | ||
XM_015135409.2 | XP_014990895.1 | 994 | 132 | |||
XM_028846756.1 | XP_028702589.1 | 1003 | 132 | |||
GenBank mRNA | JU323205.1 | - | 784 | 132 | ||
JU474606.1 | - | 785 | 132 | |||
JV044715.1 | - | 784 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |