Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260957.1 | NP_001247886.1 | 461 | 126 | ||
XM_015141101.2 | XP_014996587.1 | 747 | 126 | |||
GenBank mRNA | JU332312.1 | - | 401 | 126 | ||
JU332313.1 | - | 401 | 126 | |||
JU332314.1 | - | 401 | 126 | |||
JU332315.1 | - | 401 | 126 | |||
JU332316.1 | - | 401 | 126 | |||
JU472191.1 | - | 422 | 126 | |||
JV045670.1 | - | 461 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |