Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194300.2 | NP_001181229.1 | 368 | 110 | ||
XM_015138967.2 | XP_014994453.1 | 420 | 110 | |||
GenBank mRNA | JV072776.1 | - | 106 | 110 | ||
JV635675.1 | - | 371 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |