Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXDNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145355.2 | XP_015000841.2 | 5234 | 137 | ||
XM_015145356.2 | XP_015000842.2 | 3903 | 137 | |||
XM_015145357.2 | XP_015000843.2 | 4967 | 137 | |||
XM_015145358.2 | XP_015000844.2 | 2825 | 137 | |||
XM_015145359.2 | XP_015000845.2 | 3184 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |