Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM76B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193920.1 | NP_001180849.1 | 682 | 76 | ||
XM_015115455.2 | XP_014970941.1 | 786 | 76 | |||
GenBank mRNA | JU322146.1 | - | 380 | 76 | ||
JU473246.1 | - | 514 | 76 | |||
JV707417.1 | - | 349 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |