Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118605.2 | XP_014974091.1 | 1098 | 145 | ||
XM_015118606.2 | XP_014974092.1 | 913 | 145 | |||
XM_015118607.2 | XP_014974093.1 | 1494 | 145 | |||
XM_015118608.2 | XP_014974094.1 | 1360 | 145 | |||
GenBank mRNA | JU336277.1 | - | 819 | 145 | ||
JU477119.1 | - | 844 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |