Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOSPHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194769.2 | NP_001181698.1 | 280 | 265 | ||
XM_015110302.2 | XP_014965788.1 | 321 | 265 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |