Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114109.1 | NP_001107581.1 | 2261 | 165 | ||
XM_028838407.1 | XP_028694240.1 | 2237 | 165 | |||
GenBank mRNA | EU340327.1 | - | 2261 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |