Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091617.4 | XP_001091617.3 | 1632 | 106 | ||
XM_015121973.2 | XP_014977459.1 | 1637 | 106 | |||
XM_015121974.2 | XP_014977460.1 | 1637 | 106 | |||
GenBank mRNA | JV645215.1 | - | 349 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |