Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001088741.4 | XP_001088741.2 | 410 | 114 | ||
XM_015144362.2 | XP_014999848.1 | 410 | 114 | |||
XM_015144363.2 | XP_014999849.1 | 410 | 114 | |||
GenBank mRNA | JU336988.1 | - | 299 | 114 | ||
JU473775.1 | - | 269 | 114 | |||
JV043315.1 | - | 269 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |