Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266720.2 | NP_001253649.1 | 57 | 97 | ||
GenBank mRNA | JU322797.1 | - | 161 | 97 | ||
JU472827.1 | - | 150 | 97 | |||
JV045627.1 | - | 158 | 97 | |||
JV565064.1 | - | 57 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |