Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102810.4 | XP_001102810.3 | 782 | 107 | ||
XM_015151690.2 | XP_015007176.2 | 722 | 107 | |||
XM_015151691.2 | XP_015007177.2 | 722 | 107 | |||
XM_015151692.2 | XP_015007178.1 | 722 | 107 | |||
XM_028829467.1 | XP_028685300.1 | 722 | 107 | |||
XM_028829468.1 | XP_028685301.1 | 722 | 107 | |||
GenBank mRNA | JU337024.1 | - | 509 | 107 | ||
JV075212.1 | - | 442 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |