Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266136.1 | NP_001253065.1 | 1373 | 136 | ||
XM_015137255.2 | XP_014992741.2 | 1811 | 136 | |||
XM_015137256.2 | XP_014992742.1 | 1776 | 136 | |||
GenBank mRNA | JU474061.1 | - | 1373 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |