Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM219A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193839.2 | NP_001180768.1 | 712 | 65 | ||
XM_002800103.3 | XP_002800149.1 | 696 | 65 | |||
GenBank mRNA | JU322134.1 | - | 665 | 65 | ||
JU322135.1 | - | 665 | 65 | |||
JU322219.1 | - | 716 | 65 | |||
JU472014.1 | - | 648 | 65 | |||
JV601564.1 | - | 716 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |