Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT61A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144519.2 | XP_015000005.1 | 141 | 118 | ||
GenBank mRNA | JV044583.1 | - | 93 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |