Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087954.4 | XP_001087954.1 | 144 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |