Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194359.1 | NP_001181288.1 | 794 | 111 | ||
GenBank mRNA | JU323478.1 | - | 610 | 111 | ||
JV495664.1 | - | 679 | 111 | |||
JV534235.1 | - | 629 | 111 | |||
JV600241.1 | - | 644 | 111 | |||
JV643048.1 | - | 768 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |