Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115911.4 | XP_001115911.3 | 1247 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |