Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086115.4 | XP_001086115.2 | 1252 | 144 | ||
XM_001086220.4 | XP_001086220.2 | 1252 | 144 | |||
XM_015146430.2 | XP_015001916.2 | 1252 | 144 | |||
XM_015146447.2 | XP_015001933.2 | 1252 | 144 | |||
XM_015146461.2 | XP_015001947.2 | 1252 | 144 | |||
XM_028830849.1 | XP_028686682.1 | 1247 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |