Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194393.1 | NP_001181322.1 | 138 | 72 | ||
GenBank mRNA | JV647379.1 | - | 76 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |