Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112318.2 | XP_014967804.1 | 768 | 88 | ||
XM_015112321.2 | XP_014967807.1 | 804 | 88 | |||
XM_028831687.1 | XP_028687520.1 | 617 | 88 | |||
XM_028831688.1 | XP_028687521.1 | 751 | 88 | |||
XM_028831689.1 | XP_028687522.1 | 613 | 88 | |||
XM_028831690.1 | XP_028687523.1 | 790 | 88 | |||
XM_028831691.1 | XP_028687524.1 | 557 | 88 | |||
GenBank mRNA | JU332921.1 | - | 700 | 88 | ||
JU474323.1 | - | 658 | 88 | |||
JV706950.1 | - | 722 | 88 |