Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093030.4 | XP_001093030.2 | 1050 | 97 | ||
XM_015125441.2 | XP_014980927.2 | 968 | 97 | |||
XM_015125442.2 | XP_014980928.1 | 968 | 97 | |||
XM_015125443.2 | XP_014980929.2 | 968 | 97 | |||
XM_015125444.2 | XP_014980930.2 | 524 | 97 | |||
XM_028840949.1 | XP_028696782.1 | 968 | 97 | |||
XM_028840950.1 | XP_028696783.1 | 854 | 97 | |||
XM_028840951.1 | XP_028696784.1 | 968 | 97 | |||
XM_028840952.1 | XP_028696785.1 | 968 | 97 | |||
XM_028840953.1 | XP_028696786.1 | 968 | 97 | |||
XM_028840954.1 | XP_028696787.1 | 968 | 97 | |||
XM_028840955.1 | XP_028696788.1 | 365 | 97 | |||
XM_028840956.1 | XP_028696789.1 | 616 | 97 | |||
GenBank mRNA | JU336289.1 | - | 383 | 97 | ||
JU472228.1 | - | 286 | 97 | |||
JU472229.1 | - | 429 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |