Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSMEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102600.4 | XP_001102600.1 | 143 | 126 | ||
XM_015130864.2 | XP_014986350.1 | 146 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |