Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141422.2 | XP_014996908.1 | 2708 | 131 | ||
XM_015141423.2 | XP_014996909.1 | 2722 | 131 | |||
XM_028849922.1 | XP_028705755.1 | 1662 | 131 | |||
GenBank mRNA | JU333128.1 | - | 1458 | 131 | ||
JU472458.1 | - | 1430 | 131 | |||
JV045363.1 | - | 1579 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |