Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266943.1 | NP_001253872.1 | 659 | 110 | ||
XM_028853362.1 | XP_028709195.1 | 1362 | 110 | |||
GenBank mRNA | JU327746.1 | - | 187 | 110 | ||
JV043837.1 | - | 659 | 110 | |||
JV638691.1 | - | 832 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |