Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194358.2 | NP_001181287.2 | 445 | 109 | ||
XM_015112954.2 | XP_014968440.2 | 562 | 109 | |||
GenBank mRNA | JV497533.1 | - | 72 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |