Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100303.4 | XP_001100303.3 | 749 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Macaca mulatta |