Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095830.3 | XP_001095830.1 | 800 | 136 | ||
XM_015140494.2 | XP_014995980.1 | 800 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |