Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139951.2 | XP_014995437.2 | 2853 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |