Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCMDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145491.2 | XP_015000977.2 | 412 | 111 | ||
XM_028852704.1 | XP_028708537.1 | 361 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |