Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139663.2 | XP_014995149.1 | 982 | 126 | ||
GenBank mRNA | JV644960.1 | - | 122 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |