Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113056.4 | XP_001113056.3 | 369 | 161 | ||
XM_015134461.2 | XP_014989947.2 | 371 | 161 | |||
GenBank mRNA | JU324067.1 | - | 81 | 161 | ||
JV645403.1 | - | 82 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |