Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148280.2 | XP_015003766.1 | 1525 | 134 | ||
GenBank mRNA | JU326598.1 | - | 1021 | 134 | ||
JU470953.1 | - | 1312 | 134 | |||
JV706551.1 | - | 1042 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |