Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103304.4 | XP_001103304.3 | 975 | 136 | ||
XM_015146929.2 | XP_015002415.2 | 1311 | 136 | |||
XM_015146930.2 | XP_015002416.2 | 1362 | 136 | |||
XM_015146931.2 | XP_015002417.2 | 1311 | 136 | |||
XM_015146933.2 | XP_015002419.2 | 1246 | 136 | |||
XM_015146934.2 | XP_015002420.2 | 876 | 136 | |||
XM_015146935.2 | XP_015002421.2 | 1286 | 136 | |||
XM_015146936.2 | XP_015002422.2 | 965 | 136 | |||
XM_028826301.1 | XP_028682134.1 | 1167 | 136 | |||
XM_028826302.1 | XP_028682135.1 | 868 | 136 | |||
GenBank mRNA | JU477874.1 | - | 1364 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |