Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112727.4 | XP_001112727.2 | 1829 | 139 | ||
XM_001112810.4 | XP_001112810.2 | 1829 | 139 | |||
XM_015143445.2 | XP_014998931.2 | 1839 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |