Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194234.2 | NP_001181163.2 | 337 | 140 | ||
XM_028831596.1 | XP_028687429.1 | 562 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |