Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133929.2 | XP_014989415.1 | 921 | 80 | ||
XM_028836515.1 | XP_028692348.1 | 1005 | 80 | |||
XM_028836522.1 | XP_028692355.1 | 1207 | 80 | |||
GenBank mRNA | JU333299.1 | - | 836 | 80 | ||
JU474612.1 | - | 783 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |