Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097523.4 | XP_001097523.1 | 608 | 102 | ||
GenBank mRNA | JU326855.1 | - | 229 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |