Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV058351.1 | - | 510 | 98 | ||
JV636105.1 | - | 391 | 98 |
Set Membership: |
![]() ![]() ![]() ![]() |